Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334878.1 | NP_001321640.1 | 6 - 7 | 2105 | 59 | |
NM_001334879.1 | NP_001321639.1 | 6 - 7 | 2100 | 59 | ||
NM_001334880.1 | NP_001321641.1 | 6 - 7 | 1463 | 59 | ||
NM_106577.4 | NP_565205.1 | 6 - 7 | 1816 | 59 | ||
GenBank mRNA | AF326910.1 | - | 6 - 7 | 1410 | 59 | |
AY113874.1 | - | 6 - 7 | 1129 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |