Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198514.1 | NP_001185443.1 | 6 - 7 | 1336 | 61 | |
NM_001334926.1 | NP_001320267.1 | 5 - 6 | 1438 | 61 | ||
NM_001334927.1 | NP_001320268.1 | 5 - 6 | 1207 | 61 | ||
NM_001334928.1 | NP_001320269.1 | 5 - 6 | 1207 | 61 | ||
NM_001334929.1 | NP_001320270.1 | 4 - 5 | 1100 | 61 | ||
NM_106633.2 | NP_565223.1 | 5 - 6 | 1262 | 61 | ||
GenBank mRNA | AF360294.1 | - | 5 - 6 | 1223 | 61 | |
AJ457046.1 | - | 5 - 6 | 1112 | 61 | ||
BT001956.1 | - | 5 - 6 | 1112 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |