Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111724.3 | NP_187502.2 | 2 - 3 | 813 | 65 | |
GenBank mRNA | AF034255.2 | - | 2 - 3 | 602 | 65 | |
KJ138834.1 | - | 2 - 3 | 578 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |