Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113672.3 | NP_189393.1 | 3 - 4 | 1042 | 117 | |
GenBank mRNA | AY058062.1 | - | 3 - 4 | 954 | 117 | |
AY090300.1 | - | 3 - 4 | 456 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |