Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PME31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113832.4 | NP_566842.1 | 5 - 6 | 950 | 71 | |
GenBank mRNA | AY070091.1 | - | 5 - 6 | 851 | 71 | |
AY084418.1 | - | 5 - 6 | 885 | 71 | ||
AY096561.1 | - | 5 - 6 | 792 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |