Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114458.4 | NP_190175.2 | 6 - 7 | 1647 | 106 | |
GenBank mRNA | AK229694.1 | - | 6 - 7 | 1544 | 106 | |
BT008583.1 | - | 6 - 7 | 1528 | 106 | ||
BT008683.1 | - | 6 - 7 | 1544 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |