Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114518.4 | NP_190235.1 | 3 - 4 | 1198 | 113 | |
GenBank mRNA | AY042791.1 | - | 3 - 4 | 979 | 113 | |
AY128813.1 | - | 3 - 4 | 840 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |