Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035744.2 | NP_001030821.1 | 6 - 7 | 1624 | 100 | |
NM_001035745.2 | NP_001030822.2 | 6 - 7 | 1291 | 100 | ||
NM_001339266.1 | NP_001326037.1 | 6 - 7 | 1401 | 100 | ||
NM_001339267.1 | NP_001326038.1 | 4 - 5 | 922 | 100 | ||
NM_114526.3 | NP_190243.2 | 6 - 7 | 1306 | 100 | ||
GenBank mRNA | AJ630475.1 | - | 5 - 6 | 930 | 100 | |
AK229705.1 | - | 6 - 7 | 1213 | 100 | ||
AY395985.1 | - | 6 - 7 | 1262 | 100 | ||
AY519541.1 | - | 5 - 6 | 930 | 100 | ||
AY568647.1 | - | 5 - 6 | 930 | 100 | ||
BT033142.1 | - | 5 - 6 | 933 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |