Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114564.3 | NP_190281.1 | 3 - 4 | 793 | 64 | |
GenBank mRNA | AY090236.1 | - | 3 - 4 | 514 | 64 | |
BT003012.1 | - | 2 - 3 | 382 | 64 | ||
KJ138797.1 | - | 2 - 3 | 382 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |