Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340684.1 | NP_001328829.1 | 7 - 8 | 1886 | 123 | |
NM_117145.3 | NP_192815.1 | 8 - 9 | 1847 | 123 | ||
GenBank mRNA | AY045668.1 | - | 8 - 9 | 1771 | 123 | |
BT004519.1 | - | 8 - 9 | 1688 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |