Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203837.1 | NP_001190766.1 | 6 - 7 | 1056 | 136 | |
NM_001341309.1 | NP_001328877.1 | 6 - 7 | 1380 | 136 | ||
NM_001341310.1 | NP_001328876.1 | 6 - 7 | 1056 | 136 | ||
NM_118013.2 | NP_567569.3 | 5 - 6 | 974 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |