Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118041.5 | NP_193656.2 | 5 - 6 | 608 | 78 | |
GenBank mRNA | AK226939.1 | - | 5 - 6 | 515 | 78 | |
AY099697.1 | - | 5 - 6 | 528 | 78 | ||
BT000298.1 | - | 4 - 5 | 404 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |