Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QQT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125789.1 | NP_001119261.1 | 8 - 9 | 738 | 155 | |
NM_001343725.1 | NP_001332350.1 | 7 - 8 | 1001 | 155 | ||
NM_122142.4 | NP_197629.2 | 8 - 9 | 808 | 155 | ||
GenBank mRNA | AK221541.1 | - | 8 - 9 | 707 | 155 | |
AX765846.1 | - | 7 - 8 | 636 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |