Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122225.3 | NP_197710.1 | 1 - 2 | 1570 | 78 | |
GenBank mRNA | AF345898.1 | - | 1 - 2 | 1435 | 78 | |
BT002045.1 | - | 1 - 2 | 1286 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |