Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203480.1 | NP_001190409.1 | 5 - 6 | 854 | 160 | |
NM_122585.5 | NP_198055.3 | 5 - 6 | 1012 | 160 | ||
GenBank mRNA | BT004037.1 | - | 5 - 6 | 843 | 160 | |
BT005165.1 | - | 5 - 6 | 710 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |