Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTC-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123861.1 | NP_001117333.1 | 1 - 2 | 212 | 83 | |
NM_102130.4 | NP_173697.1 | 1 - 2 | 212 | 83 | ||
GenBank mRNA | AY062853.1 | - | 1 - 2 | 202 | 83 | |
AY087108.1 | - | 1 - 2 | 209 | 83 | ||
AY114580.1 | - | 1 - 2 | 115 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |