Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102204.3 | NP_173769.1 | 3 - 4 | 1040 | 131 | |
GenBank mRNA | BT022003.1 | - | 3 - 4 | 966 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |