Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332620.1 | NP_001321762.1 | 6 - 7 | 1003 | 76 | |
NM_102221.5 | NP_173785.1 | 7 - 8 | 641 | 76 | ||
GenBank mRNA | AY081265.1 | - | 7 - 8 | 574 | 76 | |
AY114551.1 | - | 6 - 7 | 504 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |