Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198151.2 | NP_001185080.1 | 11 - 12 | 1687 | 108 | |
NM_001332641.1 | NP_001323398.1 | 11 - 12 | 1643 | 108 | ||
NM_102265.3 | NP_173829.3 | 11 - 12 | 1624 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |