Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344352.1 | NP_001330361.1 | 14 - 15 | 1550 | 82 | |
NM_123441.3 | NP_198892.1 | 14 - 15 | 1615 | 82 | ||
GenBank mRNA | AK227918.1 | - | 14 - 15 | 1530 | 82 | |
BT004633.1 | - | 13 - 14 | 1337 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |