Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UK/UPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123452.5 | NP_198903.1 | 11 - 12 | 1372 | 76 | |
GenBank mRNA | AY089970.1 | - | 11 - 12 | 1210 | 76 | |
BT002336.1 | - | 11 - 12 | 1150 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |