Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123486.4 | NP_198937.1 | 4 - 5 | 431 | 72 | |
GenBank mRNA | AY054659.1 | - | 4 - 5 | 270 | 72 | |
AY072466.1 | - | 4 - 5 | 256 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |