Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344444.1 | NP_001318731.1 | 12 - 13 | 3352 | 108 | |
NM_001344445.1 | NP_001330664.1 | 12 - 13 | 3354 | 108 | ||
NM_001344446.1 | NP_001330665.1 | 12 - 13 | 3343 | 108 | ||
NM_001344447.1 | NP_001330668.1 | 12 - 13 | 3351 | 108 | ||
NM_001344448.1 | NP_001330669.1 | 12 - 13 | 3351 | 108 | ||
NM_001344449.1 | NP_001330666.1 | 12 - 13 | 3352 | 108 | ||
NM_001344450.1 | NP_001330667.1 | 11 - 12 | 3174 | 108 | ||
NM_001344451.1 | NP_001330670.1 | 12 - 13 | 3350 | 108 | ||
GenBank mRNA | EU014691.1 | - | 11 - 12 | 3143 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |