Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036937.1 | NP_001032014.1 | 6 - 7 | 1372 | 71 | |
NM_001344606.1 | NP_001330613.1 | 6 - 7 | 1461 | 71 | ||
NM_123898.3 | NP_199343.1 | 6 - 7 | 1376 | 71 | ||
GenBank mRNA | AK117140.1 | - | 6 - 7 | 1373 | 71 | |
BT006482.1 | - | 6 - 7 | 1350 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |