Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123958.3 | NP_568652.1 | 3 - 4 | 731 | 104 | |
GenBank mRNA | AB073173.1 | - | 3 - 4 | 684 | 104 | |
AF492761.1 | - | 3 - 4 | 602 | 104 | ||
AY034945.1 | - | 3 - 4 | 662 | 104 | ||
AY150456.1 | - | 3 - 4 | 602 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |