Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123990.3 | NP_568658.1 | 10 - 11 | 1855 | 140 | |
GenBank mRNA | AY084267.1 | - | 10 - 11 | 1714 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |