Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124045.5 | NP_199487.1 | 3 - 4 | 771 | 88 | |
GenBank mRNA | AY099693.1 | - | 3 - 4 | 695 | 88 | |
AY128872.1 | - | 3 - 4 | 607 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |