Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124091.5 | NP_568678.1 | 4 - 5 | 359 | 71 | |
GenBank mRNA | AF324990.2 | - | 4 - 5 | 309 | 71 | |
AK227103.1 | - | 4 - 5 | 322 | 71 | ||
AY080753.1 | - | 4 - 5 | 288 | 71 | ||
AY085384.1 | - | 4 - 5 | 313 | 71 | ||
AY114013.1 | - | 3 - 4 | 189 | 71 | ||
BT023739.1 | - | 3 - 4 | 189 | 71 | ||
BT025777.1 | - | 3 - 4 | 189 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |