Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124177.4 | NP_568690.1 | 4 - 5 | 539 | 147 | |
GenBank mRNA | AK118588.1 | - | 4 - 5 | 446 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |