Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344934.1 | NP_001332209.1 | 2 - 3 | 217 | 82 | |
NM_124512.3 | NP_568757.2 | 3 - 4 | 219 | 82 | ||
GenBank mRNA | AF362001.1 | - | 2 - 3 | 176 | 82 | |
AF466153.1 | - | 3 - 4 | 218 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |