Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129478.5 | NP_565902.1 | 9 - 10 | 1056 | 83 | |
GenBank mRNA | AF369573.1 | - | 9 - 10 | 926 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |