Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGGT-I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129513.4 | NP_181487.1 | 10 - 11 | 1074 | 112 | |
GenBank mRNA | BT002465.1 | - | 10 - 11 | 1022 | 112 | |
BT008464.1 | - | 10 - 11 | 979 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |