Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129693.4 | NP_181661.1 | 1 - 2 | 551 | 61 | |
GenBank mRNA | BT015732.1 | - | 1 - 2 | 317 | 61 | |
BT020191.1 | - | 1 - 2 | 276 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |