Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPB5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336901.1 | NP_001324726.1 | 2 - 3 | 709 | 81 | |
NM_001336902.1 | NP_001324725.1 | 2 - 3 | 682 | 81 | ||
NM_129697.3 | NP_181665.1 | 2 - 3 | 682 | 81 | ||
GenBank mRNA | AY078926.1 | - | 2 - 3 | 434 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |