Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFIIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129726.4 | NP_181694.1 | 5 - 6 | 947 | 136 | |
GenBank mRNA | AY086647.1 | - | 5 - 6 | 923 | 136 | |
AY093060.1 | - | 5 - 6 | 923 | 136 | ||
AY128769.1 | - | 5 - 6 | 725 | 136 | ||
AY463600.1 | - | 5 - 6 | 725 | 136 | ||
U31096.1 | - | 5 - 6 | 725 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |