Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129750.4 | NP_181717.3 | 6 - 7 | 1809 | 109 | |
NM_201932.2 | NP_973661.1 | 6 - 7 | 1438 | 109 | ||
GenBank mRNA | DQ446617.1 | - | 6 - 7 | 1438 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |