Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334708.1 | NP_001320822.1 | 1 - 2 | 946 | 145 | |
NM_202421.3 | NP_974150.2 | 1 - 2 | 946 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |