Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPE3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084425.2 | NP_001077894.1 | 1 - 2 | 572 | 149 | |
NM_127100.4 | NP_179142.1 | 1 - 2 | 572 | 149 | ||
GenBank mRNA | AK228411.1 | - | 1 - 2 | 531 | 149 | |
BT005715.1 | - | 1 - 2 | 531 | 149 | ||
BT006072.1 | - | 1 - 2 | 445 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |