Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343878.1 | NP_001318637.1 | 1 - 2 | 1143 | 108 | |
NM_180738.3 | NP_851069.1 | 1 - 2 | 1143 | 108 | ||
GenBank mRNA | AK227211.1 | - | 1 - 2 | 1143 | 108 | |
BT020231.1 | - | 1 - 2 | 1083 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |