Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103153.2 | NP_174691.2 | 12 - 13 | 1545 | 163 | |
GenBank mRNA | AB493494.1 | - | 12 - 13 | 1545 | 163 | |
AY669792.1 | - | 12 - 13 | 1545 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |