Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103250.1 | NP_174786.1 | 10 - 11 | 994 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |