Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GA20OX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104778.1 | NP_176294.1 | 1 - 2 | 550 | 151 | |
GenBank mRNA | DQ056503.1 | - | 1 - 2 | 550 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |