Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340028.1 | NP_001327770.1 | 3 - 4 | 2499 | 147 | |
NM_001340029.1 | NP_001327769.1 | 2 - 3 | 2845 | 147 | ||
NM_115879.7 | NP_191575.2 | 3 - 4 | 2678 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |