Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341319.1 | NP_001328744.1 | 3 - 4 | 925 | 135 | |
NM_118021.4 | NP_567572.1 | 4 - 5 | 774 | 135 | ||
GenBank mRNA | AY063796.1 | - | 4 - 5 | 753 | 135 | |
AY117197.1 | - | 4 - 5 | 675 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |