Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203913.1 | NP_001190842.1 | 5 - 6 | 1215 | 133 | |
NM_118718.2 | NP_567732.1 | 5 - 6 | 1014 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |