Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085300.1 | NP_001078769.1 | 3 - 4 | 1439 | 146 | |
NM_125328.4 | NP_200745.1 | 4 - 5 | 1306 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |