Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103048.5 | NP_564418.1 | 3 | 940 | 106 |
Gene Symbol: AT1G33120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF375419.1 | - | 3 | 853 | 106 | |
AY039593.1 | - | 3 | 884 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |