Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOPP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180754.4 | NP_851085.1 | 3 - 4 | 1283 | 91 | |
GenBank mRNA | AY042854.1 | - | 3 - 4 | 1142 | 91 | |
AY081492.1 | - | 3 - 4 | 939 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |