Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001600.2 | NP_001001600.2 | 3 - 4 | 367 | 73 | |
GenBank mRNA | AY330769.1 | - | 2 - 3 | 288 | 73 | |
BC149644.1 | - | 2 - 3 | 317 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |