Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013003.1 | NP_001013021.1 | 4 - 5 | 722 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Bos taurus Probe spans exons |